Boc-n-Me-abz-OH 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR960977-1G OR960977-5G
APOSOR960977-1GEA 118 GBP
APOSOR960977-1G APOSOR960977-5G
Boc-n-Me-abz-OH 97%
Boc-n-Me-abz-OH
Formula: C₁₃H₁₇NO₄
Storage Temperature: Ambient
MDL Number: MFCD00237567
CAS Number: 141871-02-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR