1-Isoxazol-5-yl-ethylamine hydrochloride 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR960585-100MG
APOSOR960585-100MGEA 210 GBP
APOSOR960585-100MG
1-Isoxazol-5-yl-ethylamine hydrochloride 95%
1-Isoxazol-5-yl-ethylamine hydrochloride
Formula: C₅H₈N₂O·ClH
Storage Temperature: Ambient
MDL Number: MFCD06738810
CAS Number: 1187930-08-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR