2-Oxa-6-azaspiro[3.3]heptane-6-ethanamine

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR959446-1G
APOSOR959446-1GEA 829 GBP
APOSOR959446-1G
2-Oxa-6-azaspiro[3.3]heptane-6-ethanamine
2-Oxa-6-azaspiro[3.3]heptane-6-ethanamine
Formula: C₇H₁₄N₂O
Storage Temperature: Ambient
MDL Number: MFCD13184501
CAS Number: 54384-40-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR