Methyl-5-aminopicolinate 96%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR959381-10G OR959381-1G OR959381-5G
APOSOR959381-10GEA 179 GBP
APOSOR959381-10G APOSOR959381-1G APOSOR959381-5G
Methyl-5-aminopicolinate 96%
Methyl 5-aminopicolinate
Formula: C₇H₈N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD07783939
CAS Number: 67515-76-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR