N-(tert-Butoxycarbonyl)-D-threonine methyl ester 98+%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Boc-D-Thr-OMe, N-Boc-D-threonine methyl ester, (2R,3S)-Methyl 2-((tert-butoxycarbonyl)amino)-3-hydroxybutanoate

OR955606-1G OR955606-5G
APOSOR955606-1GEA 211 GBP
APOSOR955606-1G APOSOR955606-5G
N-(tert-Butoxycarbonyl)-D-threonine methyl ester 98+%
N-(tert-Butoxycarbonyl)-D-threonine methyl ester
Formula: C₁₀H₁₉NO₅
MW: 233.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01073763
CAS Number: 96099-84-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR