Ethylene glycol monoallyl ether 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 2-(Allyloxy)ethanol, 2-Allyloxyethanol, 2-(2-Propenyloxy)ethanol, Allyl glycol

OR955005-100G OR955005-1KG OR955005-500G
APOSOR955005-100GEA 89.8 GBP
APOSOR955005-100G APOSOR955005-1KG APOSOR955005-500G
Ethylene glycol monoallyl ether 95%
Ethylene glycol monoallyl ether
Formula: C₅H₁₀O₂
Storage Temperature: Ambient
MDL Number: MFCD00020607
CAS Number: 111-45-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR