DL-Menthol 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR954314-100G
APOSOR954314-100GEA 31.5 GBP
APOSOR954314-100G
DL-Menthol 95%
DL-Menthol
Formula: C₁₀H₂₀O
Storage Temperature: Ambient
MDL Number: MFCD00001484
CAS Number: 89-78-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR