Methyl 3-methyl-2-oxobutanoate 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR952085-1G OR952085-250MG
APOSOR952085-1GEA 415 GBP
APOSOR952085-1G APOSOR952085-250MG
Methyl 3-methyl-2-oxobutanoate 95%
Methyl 3-methyl-2-oxobutanoate
Formula: C₆H₁₀O₃
Storage Temperature: Ambient
MDL Number: MFCD12198846
CAS Number: 3952-67-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR