6-Azaspiro[2.5]octan-4-ol hydrochloride

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR952020-1G
APOSOR952020-1GEA 932 GBP
APOSOR952020-1G
6-Azaspiro[2.5]octan-4-ol hydrochloride
6-Azaspiro[2.5]octan-4-ol hydrochloride
Formula: C₇H₁₃NO·ClH
MW: 163.645 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21607169
CAS Number: 955028-68-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR