D-N-Benzylserine methyl ester 96%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: N-Benzyl-D-Serine-OMe Ester, Methyl (2R)-2-(benzylamino)-3-hydroxypropanoate, D-N-Benzylserine Methyl Ester

OR951335-1G OR951335-5G
APOSOR951335-1GEA 115 GBP
APOSOR951335-1G APOSOR951335-5G
D-N-Benzylserine methyl ester 96%
D-N-Benzylserine methyl ester
Formula: C₁₁H₁₅NO₃
Storage Temperature: Freezer
MDL Number: MFCD12198843
CAS Number: 131110-76-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR