Diisopropyl fumarate 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR950492-1G OR950492-5G
APOSOR950492-1GEA 15.8 GBP
APOSOR950492-1G APOSOR950492-5G
Diisopropyl fumarate 98%
Diisopropyl fumarate
Formula: C₁₀H₁₆O₄
Storage Temperature: Ambient
MDL Number: MFCD00545172
CAS Number: 7283-70-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR