3-Nitro-2,6-lutidine

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

OR9504-25G OR9504-5G OR9504-100G
APOSOR9504-25GEA 53.6 GBP
APOSOR9504-25G APOSOR9504-5G APOSOR9504-100G
3-Nitro-2,6-lutidine
3-Nitro-2,6-lutidine
Formula: C₇H₈N₂O₂
Melting Pt: 26…31 °C
Flash Pt: 37 °C
Storage Temperature: Ambient
MDL Number: MFCD00023503
CAS Number: 15513-52-7
EINECS: 239-543-5
UN: 1325
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR