2-(Aminomethyl)pyridin-3-ol 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR950277-250MG
APOSOR950277-250MGEA 141 GBP
APOSOR950277-250MG
2-(Aminomethyl)pyridin-3-ol 97%
2-(Aminomethyl)pyridin-3-ol
Formula: C₆H₈N₂O
MW: 124.141 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01217253
CAS Number: 194665-89-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR