5-Methyl-2-thenoic acid

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 5-Methylthiophene-2-carboxylic acid

OR9459-100G OR9459-25G OR9459-5G
APOSOR9459-100GEA 79.2 GBP
APOSOR9459-100G APOSOR9459-25G APOSOR9459-5G
5-Methyl-2-thenoic acid
5-Methyl-2-thenoic acid
Formula: C₆H₆O₂S
Melting Pt: 135 °C
Storage Temperature: Ambient
MDL Number: MFCD00005439
CAS Number: 1918-79-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR