(Tetrahydro-pyran-2-yloxy)-acetonitrile 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR944255-1G
APOSOR944255-1GEA 405 GBP
APOSOR944255-1G
(Tetrahydro-pyran-2-yloxy)-acetonitrile 95%
(Tetrahydro-pyran-2-yloxy)-acetonitrile
Formula: C₇H₁₁NO₂
Storage Temperature: Ambient
MDL Number: MFCD21362389
CAS Number: 17521-49-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR