5-Chloro-2-(methylamino)phenol 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR942472-1G OR942472-250MG
APOSOR942472-1GEA 415 GBP
APOSOR942472-1G APOSOR942472-250MG
5-Chloro-2-(methylamino)phenol 95%
5-Chloro-2-(methylamino)phenol
Formula: C₇H₈ClNO
Storage Temperature: Ambient
MDL Number: MFCD16998610
CAS Number: 1243357-05-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR