3-(Methylamino)phenol 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR941589-1G OR941589-250MG
APOSOR941589-1GEA 95.7 GBP
APOSOR941589-1G APOSOR941589-250MG
3-(Methylamino)phenol 97%
3-(Methylamino)phenol
Formula: C₇H₉NO
Storage Temperature: Ambient
MDL Number: MFCD16998628
CAS Number: 14703-69-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR