2,5-Dimethyl-4-nitrobenzonitrile 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR939982-1G OR939982-250MG OR939982-500MG
APOSOR939982-1GEA 504 GBP
APOSOR939982-1G APOSOR939982-250MG APOSOR939982-500MG
2,5-Dimethyl-4-nitrobenzonitrile 95%
2,5-Dimethyl-4-nitrobenzonitrile
Formula: C₉H₈N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00024285
CAS Number: 73713-69-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR