Ethyl 2-bromoacrylate 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR939045-1G OR939045-5G
APOSOR939045-1GEA 189 GBP
APOSOR939045-1G APOSOR939045-5G
Ethyl 2-bromoacrylate 95%
Ethyl 2-bromoacrylate
Formula: C₅H₇BrO₂
Storage Temperature: Ambient
MDL Number: MFCD02258596
CAS Number: 5459-35-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR