2-Methoxy-1-butanol 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR937857-10G OR937857-1G OR937857-5G
APOSOR937857-10GEA 468 GBP
APOSOR937857-10G APOSOR937857-1G APOSOR937857-5G
2-Methoxy-1-butanol 95%
2-Methoxy-1-butanol
Formula: C₅H₁₂O₂
Storage Temperature: Ambient
CAS Number: 15467-25-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR