3-(4-Methoxyphenyl)propionyl chloride 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 3-(4-Methoxyphenyl)propanoyl chloride

OR937079-1G OR937079-5G
APOSOR937079-1GEA 44.1 GBP
APOSOR937079-1G APOSOR937079-5G
3-(4-Methoxyphenyl)propionyl chloride 95%
3-(4-Methoxyphenyl)propionyl chloride
Formula: C₁₀H₁₁ClO₂
Storage Temperature: Ambient
MDL Number: MFCD02258671
CAS Number: 15893-42-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR