(S)-(+)-3-Methyl-1-pentanol 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR937032-1G OR937032-250MG
APOSOR937032-1GEA 289 GBP
APOSOR937032-1G APOSOR937032-250MG
(S)-(+)-3-Methyl-1-pentanol 95%
(S)-(+)-3-Methyl-1-pentanol
Formula: C₆H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD00135167
CAS Number: 42072-39-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR