Hexamethyldisiloxane 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Silicone fluid, HMDSO

OR936705-100G
APOSOR936705-100GEA 89.8 GBP
APOSOR936705-100G
Hexamethyldisiloxane 95%
Hexamethyldisiloxane
Formula: C₆H₁₈OSi₂
MW: 162.38 g/mol
Boiling Pt: 101
Melting Pt: -59
Flash Pt: 0.6
Storage Temperature: Ambient
MDL Number: MFCD00008265
CAS Number: 107-46-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR