4-Methyl-1,2-cyclohexanedicarboxylic anhydride (mixture of cis- and trans-isomeres) 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Hexahydro-4-methylphthalic anhydride

OR936561-100G OR936561-1KG OR936561-500G
APOSOR936561-100GEA 89.8 GBP
APOSOR936561-100G APOSOR936561-1KG APOSOR936561-500G
4-Methyl-1,2-cyclohexanedicarboxylic anhydride (mixture of cis- and trans-isomeres) 95%
4-Methyl-1,2-cyclohexanedicarboxylic anhydride (mixture of cis- and trans-isomeres)
Formula: C₉H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00005927
CAS Number: 19438-60-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR