2-Phenylpropionitrile 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: alpha-Methylphenylacetonitrile

OR936420-25G
APOSOR936420-25GEA 155 GBP
APOSOR936420-25G
2-Phenylpropionitrile 95%
2-Phenylpropionitrile
Formula: C₉H₉N
MW: 131.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00001871
CAS Number: 1823-91-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR