N-Methyl-DL-alanine

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Me-DL-Ala-OH, H-N-Me-DL-Ala-OH, N-DL-Me-Ala-OH

OR930477-100G OR930477-1KG OR930477-500G
APOSOR930477-100GEA 89.8 GBP
APOSOR930477-100G APOSOR930477-1KG APOSOR930477-500G
N-Methyl-DL-alanine
N-Methyl-DL-alanine
Formula: C₄H₉NO₂
Melting Pt: 317 °C
Storage Temperature: Ambient
MDL Number: MFCD00063136
CAS Number: 600-21-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR