1-Benzyl-azetidine-2-carboxylic acid amide 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 1-Benzylazetidine-2-carboxamide

OR927776-1G OR927776-250MG
APOSOR927776-1GEA 357 GBP
APOSOR927776-1G APOSOR927776-250MG
1-Benzyl-azetidine-2-carboxylic acid amide 95%
1-Benzyl-azetidine-2-carboxylic acid amide
Formula: C₁₁H₁₄N₂O
Storage Temperature: Ambient
MDL Number: MFCD06658337
CAS Number: 40432-40-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR