tert-Butyl-2-hydroxy-2-methylpropylcarbamate 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR927648-1G
APOSOR927648-1GEA 26.3 GBP
APOSOR927648-1G
tert-Butyl-2-hydroxy-2-methylpropylcarbamate 95%
tert-Butyl-2-hydroxy-2-methylpropylcarbamate
Formula: C₉H₁₉NO₃
Storage Temperature: Ambient
MDL Number: MFCD09608009
CAS Number: 183059-24-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR