2,5-Dichloroquinoxaline 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR927312-1G OR927312-250MG
APOSOR927312-1GEA 47.3 GBP
APOSOR927312-1G APOSOR927312-250MG
2,5-Dichloroquinoxaline 95%
2,5-Dichloroquinoxaline
Formula: C₈H₄Cl₂N₂
Storage Temperature: Ambient
MDL Number: MFCD11846477
CAS Number: 55687-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR