4'-Chloro-2'-nitroacetophenone 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR926954-5G
APOSOR926954-5GEA 188 GBP
APOSOR926954-5G
4'-Chloro-2'-nitroacetophenone 98%
4'-Chloro-2'-nitroacetophenone
Formula: C₈H₆ClNO₃
Storage Temperature: Ambient
MDL Number: MFCD09031331
CAS Number: 23082-51-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR