5-Methoxy-1H-pyrrolo[2,3-c]pyridine-2-carboxylic acid 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR926791-100MG OR926791-1G OR926791-250MG
APOSOR926791-100MGEA 83.9 GBP
APOSOR926791-100MG APOSOR926791-1G APOSOR926791-250MG
5-Methoxy-1H-pyrrolo[2,3-c]pyridine-2-carboxylic acid 95%
5-Methoxy-1H-pyrrolo[2,3-c]pyridine-2-carboxylic acid
Formula: C₉H₈N₂O₃
Storage Temperature: Ambient
MDL Number: MFCD08448208
CAS Number: 17288-36-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR