2-Methylthiazole-5-methanol 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR925922-1G OR925922-250MG OR925922-5G
APOSOR925922-1GEA 130 GBP
APOSOR925922-1G APOSOR925922-250MG APOSOR925922-5G
2-Methylthiazole-5-methanol 95%
2-Methylthiazole-5-methanol
Formula: C₅H₇NOS
Storage Temperature: Ambient
MDL Number: MFCD12912988
CAS Number: 56012-38-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR