Lead tetraacetate 96%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Lead(IV) tetraacetate, Triacetyloxyplumbyl acetate, Acetic acid lead (IV) salt, Lead (IV) acetate, LTA

OR924077-5G OR924077-25G OR924077-100G
APOSOR924077-5GEA 15.8 GBP
APOSOR924077-5G APOSOR924077-25G APOSOR924077-100G
Lead tetraacetate 96%
Lead tetraacetate
Formula: (H₃CCOO)₄Pb
MW: 443.38 g/mol
Melting Pt: 175…180 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008693
CAS Number: 546-67-8
UN: 1616
ADR: 6.1 III
Merck Index: 13,05442

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR