2,7-Diazaspiro[4.4]nonan-1-one hydrochloride 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR922643-1G OR922643-500MG
APOSOR922643-1GEA 499 GBP
APOSOR922643-1G APOSOR922643-500MG
2,7-Diazaspiro[4.4]nonan-1-one hydrochloride 97%
2,7-Diazaspiro[4.4]nonan-1-one hydrochloride
Formula: C₇H₁₂N₂O·ClH
Storage Temperature: Refrigerator
MDL Number: MFCD10700079
CAS Number: 1203682-17-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR