Ethylene glycol monoisopropyl ether 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 2-Isopropoxyethanol

OR922073-100G OR922073-1KG OR922073-500G
APOSOR922073-100GEA 78.7 GBP
APOSOR922073-100G APOSOR922073-1KG APOSOR922073-500G
Ethylene glycol monoisopropyl ether 95%
Ethylene glycol monoisopropyl ether
Formula: C₅H₁₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00002866
CAS Number: 109-59-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR