N-Ethyl-1-methylazetidin-3-amine

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR921221-1G
APOSOR921221-1GEA 603 GBP
APOSOR921221-1G
N-Ethyl-1-methylazetidin-3-amine
N-Ethyl-1-methylazetidin-3-amine
Formula: C₆H₁₄N₂
MW: 114.189 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24335256
CAS Number: 1434128-51-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR