3-Bromo-1-methyl-1H-1,2,4-triazole 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR917584-5G
APOSOR917584-5GEA 49.4 GBP
APOSOR917584-5G
3-Bromo-1-methyl-1H-1,2,4-triazole 97%
3-Bromo-1-methyl-1H-1,2,4-triazole
Formula: C3H4BrN3
MW: 161.9900 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11109755
CAS Number: 56616-91-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR