2,5-Dichloroquinoline 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR917331-1G
APOSOR917331-1GEA 42 GBP
APOSOR917331-1G
2,5-Dichloroquinoline 95%
2,5-Dichloroquinoline
Formula: C₉H₅Cl₂N
Storage Temperature: Ambient
MDL Number: MFCD00832074
CAS Number: 59412-12-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR