DL-Methionine methyl ester hydrochloride 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Methyl 2-amino-4-(methylthio)butanoate hydrochloride, H-DL-Met-OMe·HCl

OR916765-100G OR916765-25G OR916765-5G
APOSOR916765-100GEA 427 GBP
APOSOR916765-100G APOSOR916765-25G APOSOR916765-5G
DL-Methionine methyl ester hydrochloride 95%
DL-Methionine methyl ester hydrochloride
Formula: C₆H₁₃NO₂S·HCl
Melting Pt: 110…117 °C
Storage Temperature: Ambient
MDL Number: MFCD00067540
CAS Number: 16118-36-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR