Methyl 2-amino-6-bromobenzoate 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR916421-250MG OR916421-1G OR916421-5G
APOSOR916421-250MGEA 27.3 GBP
APOSOR916421-250MG APOSOR916421-1G APOSOR916421-5G
Methyl 2-amino-6-bromobenzoate 97%
Methyl 2-amino-6-bromobenzoate
Formula: C₈H₈BrNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD11977360
CAS Number: 135484-78-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR