2-Bromo-5-cyano-4-picoline 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR914313-5G
APOSOR914313-5GEA 170 GBP
APOSOR914313-5G
2-Bromo-5-cyano-4-picoline 95%
2-Bromo-5-cyano-4-picoline
Formula: C₇H₅BrN₂
Storage Temperature: Ambient
MDL Number: MFCD09839265
CAS Number: 1003711-35-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR