(2-Formylphenoxy)acetonitrile 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR913452-1G OR913452-5G
APOSOR913452-1GEA 44.1 GBP
APOSOR913452-1G APOSOR913452-5G
(2-Formylphenoxy)acetonitrile 97%
(2-Formylphenoxy)acetonitrile
Formula: C₉H₇NO₂
Storage Temperature: Ambient
MDL Number: MFCD01157218
CAS Number: 125418-83-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR