4-(Methylthio)-m-cresol 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 3-Methyl-4-(methylthio)phenol

OR912189-100G OR912189-25G
APOSOR912189-100GEA 252 GBP
APOSOR912189-100G APOSOR912189-25G
4-(Methylthio)-m-cresol 98%
4-(Methylthio)-m-cresol
Formula: C₈H₁₀OS
MDL Number: MFCD00045773
CAS Number: 3120-74-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR