6-Chloro-2,8-dimethylquinoline 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR911962-1G
APOSOR911962-1GEA 557 GBP
APOSOR911962-1G
6-Chloro-2,8-dimethylquinoline 95%
6-Chloro-2,8-dimethylquinoline
Formula: C₁₁H₁₀ClN
Storage Temperature: Ambient
CAS Number: 948289-20-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR