6-Nitro-o-cresol 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR911362-100G OR911362-25G
APOSOR911362-100GEA 890 GBP
APOSOR911362-100G APOSOR911362-25G
6-Nitro-o-cresol 98%
6-Nitro-o-cresol
Formula: C₇H₇NO₃
Storage Temperature: Ambient
MDL Number: MFCD00031116
CAS Number: 13073-29-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR