1-Methyl-6-nitroindole 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR910612-1G OR910612-5G
APOSOR910612-1GEA 46.2 GBP
APOSOR910612-1G APOSOR910612-5G
1-Methyl-6-nitroindole 98%
1-Methyl-6-nitroindole
Formula: C₉H₈N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD13178347
CAS Number: 99459-48-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR