1-Cyclopropyl-2,5-dimethylpyrrole 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR910266-10G OR910266-1G OR910266-5G
APOSOR910266-10GEA 363 GBP
APOSOR910266-10G APOSOR910266-1G APOSOR910266-5G
1-Cyclopropyl-2,5-dimethylpyrrole 95%
1-Cyclopropyl-2,5-dimethylpyrrole
Formula: C₉H₁₃N
Storage Temperature: Ambient
MDL Number: MFCD12187123
CAS Number: 54609-12-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR