Ethyl 5-cyclopropyl-2H-pyrazole-3-carboxylate 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR909668-5G
APOSOR909668-5GEA 139 GBP
APOSOR909668-5G
Ethyl 5-cyclopropyl-2H-pyrazole-3-carboxylate 98%
Ethyl 5-cyclopropyl-2H-pyrazole-3-carboxylate
Formula: C₉H₁₂N₂O₂
Melting Pt: 90…92 °C
Storage Temperature: Refrigerator
MDL Number: MFCD16658044
CAS Number: 133261-06-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR