2-Cyano-1,3-dihydroindene-2-carboxylic acid 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR909657-1G
APOSOR909657-1GEA 235 GBP
APOSOR909657-1G
2-Cyano-1,3-dihydroindene-2-carboxylic acid 97%
2-Cyano-1,3-dihydroindene-2-carboxylic acid
Formula: C₁₁H₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD21802888
CAS Number: 858226-43-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR