2-Acetamido-5-methylthiazole 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR909503-1G OR909503-5G
APOSOR909503-1GEA 105 GBP
APOSOR909503-1G APOSOR909503-5G
2-Acetamido-5-methylthiazole 98%
2-Acetamido-5-methylthiazole
Formula: C₆H₈N₂OS
Melting Pt: 230…232 °C
Storage Temperature: Refrigerator
MDL Number: MFCD18801061
CAS Number: 61996-32-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR